EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Rapid determination of procetofenic acid in plasma by high-performance liquid chromatography.
PMID 7056813
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0048010
Fenofibrate impurity B, European Pharmacopoeia (EP) Reference Standard
C17H15ClO4