EMAIL THIS PAGE TO A FRIEND
Mutation research
Transformation of BHK 21 C13 cells by BC, 4CMB and 4HMB using the method of Styles.
PMID 7057748
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123838
Biphenyl-4-methanol, 98%
C13H12O