EMAIL THIS PAGE TO A FRIEND
Mutation research
Induction of morphological transformation in C3H/10T1/2 clone 8 cells.
PMID 7057749
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123838
Biphenyl-4-methanol, 98%
C13H12O