EMAIL THIS PAGE TO A FRIEND
Mutation research
Investigation of the cytogenetic effects of BC and 4CMB on human peripheral lymphocytes in vitro.
PMID 7057763
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C52276
4-Phenylbenzyl chloride, 98%
C13H11Cl