EMAIL THIS PAGE TO A FRIEND
Mutation research
Cytogenetic effects of BC and 4CMB in the mouse evaluated by the micronucleus test.
PMID 7057775
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C52276
4-Phenylbenzyl chloride, 98%
C13H11Cl