EMAIL THIS PAGE TO A FRIEND
Mutation research
An evaluation of the mutagenic potential of 4-chloromethylbiphenyl (4CMB) using the micronucleus test.
PMID 7057776
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C52276
4-Phenylbenzyl chloride, 98%
C13H11Cl