EMAIL THIS PAGE TO A FRIEND
Archives of virology
Affinity chromatography of viroid RNAs.
PMID 7065904
Abstract
The application of a structure- and GC-specific affinity adsorbent for the purification of viroid RNAs is described. Cross-linked bisacrylamide gels to which the intercalating dye phenyl neutral red is covalently attached [2] constitute a powerful tool for the rapid, quantitative, and selective isolation of viroid as well as viroid complementary RNAs from crude, plant polysaccharides containing nucleic acid fractions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199508
Acridine Yellow G, Dye content 90 %
C15H15N3 · HCl