EMAIL THIS PAGE TO A FRIEND
FEBS letters
Metabolic pathway to 25-hydroxyvitamin D3-26,23-lactone from 25-hydroxyvitamin D3.
PMID 7067832
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55171
(23R)-23,25-Dihydroxyvitamin D3, ≥99.0% (HPLC)
C27H44O3