EMAIL THIS PAGE TO A FRIEND
Steroids
Acyl migration in the Reformatsky reaction of 21-acyloxy-5-pregnen-20-one derivatives with ethyl bromoacetate.
PMID 7071886
Abstract
Reformatsky reaction of 3beta, 21-diacyloxy-and 3beta-methoxy-21-acyloxy-5-pregnen-20-one with ethyl bromoacetate yields, through an intramolecular 1,2-acyl migration, 38, 20 XI-acyloxy-14alpha-card-5-enolides were converted into the respective 20 XI-hydroxy-14alpha-card-5-enolides and the 14 alpha-carda-5,20(22)-dienolides. Experimental support to the proposed intramolecular 1,2-acyl migration is provided by the use of labelled compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133973
Ethyl bromoacetate, reagent grade, 98%
C4H7BrO2
17020
Ethyl bromoacetate, purum, ≥97.0% (GC)
C4H7BrO2