EMAIL THIS PAGE TO A FRIEND
The British journal of venereal diseases
Treatment of uncomplicated gonorrhoea in women with talampicillin in a single oral 1.5 g dose.
PMID 7082979
Abstract
Four hundred and sixty women with uncomplicated gonorrhea were treated with a single oral dose of 1.5 g talampicillin. The success rate among those who attended for at least one follow-up examination after treatment was 99.14%. The antibiotic was well tolerated and no side effects were reported. Talampicillin given under supervision has proved to be the most effective and safest treatment in this area. A smaller dosage of talampicillin than ampicillin is required, the drug is better tolerated by the patient, and the time of nursing staff is saved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0010000 Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C24H23N3O6S · HCl