EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of moroxydine in biological fluids by electron-capture gas chromatography.
PMID 7085830
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
278610
Moroxydine hydrochloride, 99%
C6H13N5O · HCl