EMAIL THIS PAGE TO A FRIEND
Current surgery
Intra-abdominal sepsis and perfluorocarbons: mechanism of protection.
PMID 7094625
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
446882
Perfluorohexyl bromide, 98%
C6BrF13