EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Experimental study of the blastomogenic properties of Basic Blue K].
PMID 7095457
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234176
Victoria Blue R, Dye content 80 %
C29H32ClN3