EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Syringe method for stepwise chemical synthesis of oligonucleotides.
PMID 7099961
Abstract
A simple procedure is described for synthesis of oligonucleotides by phosphate chemistry. Chains can be constructed rapidly with minimal equipment (a syringe and reagent bottles). The method is illustrated by synthesis of d-TGCAGGTT. Pertinent supporting data on the effect of variations in the detritylation, condensation, oxidation, capping and cleavage steps in the synthetic approach and in isolation procedures are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235229
Methyl dichlorophosphite, technical grade
CH3Cl2OP