EMAIL THIS PAGE TO A FRIEND
General and comparative endocrinology
Formation of 15 alpha-hydroxylated androgens by the testis and other tissues of the sea lamprey, Petromyzon marinus, in vitro.
PMID 7106548