EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Improved method for the evaluation of 3-hydroxyproline and 4-hydroxyproline in the urine.
PMID 7107786
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420174
cis-3-Hydroxy-DL-proline, 95%
C5H9NO3