EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Ring contraction of oleandrose on the macrolide antibiotic oleandomycin with [(Methoxycarbonyl)sulfamoyl]triethylammonium hydroxide inner salt.
PMID 7108908
Abstract
Ring contraction of the neutral oleandrose sugar in the 14-membered-ring macrolide antibiotic oleandomycin (2) has been accomplished using [(methoxycarbonyl)sulfamoyl]triethylammonium hydroxide inner salt (1). The product of this interesting rearrangement, after methanolic hydrolysis of the 2'-acetate, is the 11-acetyl-3-O-(3"-methoxy-4"-vinylfuranosyl)oleandomycin (12). The in vitro activity of furanoside 12 is only moderately less than that of 11-acetyloleandomycin (13).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
365483
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt, 97%
C8H18N2O4S