EMAIL THIS PAGE TO A FRIEND
Tijdschrift voor diergeneeskunde
[Effect of and length of protection offered by Cepravin Dry Cow in the prevention of summer-mastitis (author's transl)].
PMID 7112525
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32904
Cefalonium hydrate, VETRANAL®, analytical standard
C20H18N4O5S2 · xH2O