EMAIL THIS PAGE TO A FRIEND
Zentralblatt fur Arbeitsmedizin, Arbeitsschutz, Prophylaxe und Ergonomie
[Morbidity study of employees exposed to dimethylaminopropionitrile].
PMID 7113493
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234486
3-(Dimethylamino)propionitrile, 98%
C5H10N2