EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica Suecica
Isolation and determination of the diastereomers of the glucuronidated 5-(4-hydroxyphenyl)-5-phenylhydantoin in human urine using HPLC.
PMID 7113727