EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
Isolation and mass spectrometric identification of gitoxin metabolites excreted in bile.
PMID 7117294
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64107
Gitoxin, ≥99% (HPLC)
C41H64O14