EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Cleavage of aspartyl beta-phenacyl esters by selenophenol under neutral conditions.
PMID 7118418
Abstract
Neutral selenophenol in DMF accomplished the removal of the beta-phenacyl protecting group of aspartic acid in solution, on the peptide Boc-Asp (beta-OPac)- Phe amide and on the resin peptide Boc-Trp(Nin-For)-Met-Asp (beta-OPac)-P (CCK 30-33) without alpha, beta-rearrangement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
375152
Benzeneselenol, 97%
C6H6Se