EMAIL THIS PAGE TO A FRIEND
Journal of theoretical biology
Modified nucleotides: their conformational characteristics.
PMID 7121028
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9886
2′-O-Methyladenosine
C11H15N5O4