EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Physical properties of purified calf brain tubulin.
PMID 7121278
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63064
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Cl2Mg · 6H2O