EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Photometric method of determining di(2-ethylhexyl)phenylphosphate in air].
PMID 7129149
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34064
2-Ethylhexyl diphenyl phosphate, PESTANAL®, analytical standard
C20H27O4P