EMAIL THIS PAGE TO A FRIEND
Fortschritte der Medizin
[Drug modification of a risk factor of thrombosis in patients with varicosis].
PMID 7129301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0490000 Benzarone, European Pharmacopoeia (EP) Reference Standard
C17H14O3