EMAIL THIS PAGE TO A FRIEND
Vrachebnoe delo
[Specialized outpatient care in infectious diseases].
PMID 7135894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
637602
Barium ferrite, nanopowder, <100 nm particle size (BET), >97% trace metals basis
BaFe12O19