EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Sensitisation and visualisation of biochemical measurements using the NAD/NADH system by means of Meldola blue. II. Application to the continuous flow determination of plasma glucose and urea.
PMID 7139960