EMAIL THIS PAGE TO A FRIEND
La Clinica terapeutica
[Therapeutic action of oxolamine in acute tracheo-bronchitis].
PMID 7140166
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2256
Oxolamine citrate salt
C14H19N3O · C6H8O7