EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Interference in plasma level measurements of certain drugs due to a plasticizer in Vacutainers.
PMID 7143202
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130591
Tris(2-butoxyethyl) phosphate, 94%
C18H39O7P