EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Tetramethyl substitution on succinonitrile confers pentylenetetrazol-like activity and blocks cyanide release in mice.
PMID 7143230
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160962
Succinonitrile, 99%
C4H4N2
14190
Succinonitrile, purum, ≥97.0% (GC)
C4H4N2