EMAIL THIS PAGE TO A FRIEND
Pharmaceutica acta Helvetiae
[High pressure liquid chromatography of dibenzylethylenediamine in benzathine penicillin].
PMID 7146033
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D35201
N,N′-Dibenzylethylenediamine, 97%
C16H20N2