EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Spectral and metabolic properties of liver microsomes from imidazole-pretreated rabbits.
PMID 7150315
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144754
4-Phenylimidazole, 97%
C9H8N2