EMAIL THIS PAGE TO A FRIEND
Journal of medical entomology
Tests of repellents against Diamanus montanus (Siphonaptera: Ceratophyllidae).
PMID 7154015
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E29125
2-Ethyl-1,3-hexanediol, 97%, Mixture of isomers
C8H18O2