EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Rechtsmedizin. Journal of legal medicine
[FAB (fast atom bombardment)-mass spectrometry. A new study method in the hands of the (forensic) toxicologist].
PMID 7158081
Abstract
The FAB (Fast Atom Bombardment)-mass spectrometric ionization technique, which has now been available for about 1 year, has been successfully employed in forensic toxicology. The mass spectral behaviour of some representative drug-glucuronides (Codeine, p-Nitrophenol and 2-Phenyl-1-propanol) were studied by positive- and negative-ion-FAB-MS. The presented promising results may be of some interest, not only for the analytical toxicologist.