EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Synthesis and schistosomicidal activity of fluoroacetamides--a new class of antischistosomal agents].
PMID 7158352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128341
Fluoroacetamide
C2H4FNO