EMAIL THIS PAGE TO A FRIEND
Archives of dermatological research
Allergic contact dermatitis to various salols (phenyl salicylates). A structure-activity relationship study in man and in animal (guinea pig).
PMID 7165321