EMAIL THIS PAGE TO A FRIEND
Archives of environmental contamination and toxicology
Toxicity of acenaphthene and isophorone to early life stages of fathead minnows.
PMID 7165389
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I18709
Isophorone, 97%
C9H14O
W355305
Isophorone, ≥97%, FG
C9H14O