EMAIL THIS PAGE TO A FRIEND
Cytologia
Recovery from the 5-AU induced blockage in interphase: evidence for differential recovery.
PMID 7166054
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
855286
5-Aminouracil, 98%
C4H5N3O2