EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
[Effect of simultaneous administration of drugs on absorption and excretion. XIV. Effect of atropine on gastrointestinal absorption of sulfisomidine in rabbits].
PMID 7166732
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46908
Sulfisomidin, VETRANAL, analytical standard
C12H14N4O2S