EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
A possible catalytic role of pyridoxal-5'-phosphate in a general acid base catalyzed reaction of D-glucal with glycogen phosphorylases.
PMID 7167461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464058
D-Glucal, 96%
C6H10O4