EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Use of ion cluster analysis in a metabolic study of pencycuron, a phenylurea fungicide, in rabbits.
PMID 7175035
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31118
Pencycuron, PESTANAL®, analytical standard
C19H21ClN2O