EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Inhalation toxicity of adiponitrile in rats.
PMID 7179283
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D77001
Adiponitrile, 99%
C6H8N2