EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Occurrence of aromatic methyl migration (NIH-shift) during oxidation of p-methylanisole by hemin-thiolester complex as a cytochrome P-450 model.
PMID 7181912
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148091
4-Methylanisole, 99%
C8H10O
W268100
4-Methylanisole, ≥99%, FCC, FG
C8H10O