EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
The effects of (-)N-(chloroethyl)-norapomorphine on [3H]-apomorphine binding to striatal dopamine receptors.
PMID 7181954
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D007
R(−)-Chloroethylnorapomorphine hydrochloride, analytical standard
C18H18ClNO2 · HCl