EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Phytotoxins in Rhizoctonia solani: isolation and biological activity of m-hydroxy- and m-methoxyphenylacetic acids.
PMID 7188946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M19007
3-Methoxyphenylacetic acid, ≥99%
C9H10O3