EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Microbial transformations of natural antitumor agents. 13. Conversions of 13-blocked anthracyclinones.
PMID 7190964
Abstract
Microbial transformation experiments were performed using 13-ketone-blocked daunomycinone derivatives which were converted into the corresponding carminomycinone derivatives by microbial O-demethylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0120000 Daunorubicinone, European Pharmacopoeia (EP) Reference Standard
C21H18O8