EMAIL THIS PAGE TO A FRIEND
The American journal of psychiatry
Synergism of combined lithium-neuroleptic therapy: a double-blind, placebo-controlled case study.
PMID 7192495
Abstract
The authors describe a longitudinal, placebo-controlled study of the response to drug treatment of a 62-year-old schizoaffective patient hospitalized for 40 years. While neither lithium nor a neuroleptic drug alone was effective, both drugs administered simultaneously led to the gradual extinction of a pronounced behavioral cycle and the disappearance of manifest psychosis. The authors suggest a synergistic interaction between the two drugs and stress the need for sustained trials of the combination in patients with similar behavioral cycles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1007000 Acetophenazine maleate, United States Pharmacopeia (USP) Reference Standard
C23H29N3O2S · 2C4H4O4