EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Contact dermatitis from D & C yellow 11 dye in a toilet bar soap.
PMID 7195408
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234133
Quinoline Yellow, Dye content 95 %
C18H11NO2