EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Quinazoline yellow dermatitis (D & C Yellow 11) in an eye cream.
PMID 7198023
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234133
Quinoline Yellow, Dye content 95 %
C18H11NO2